Benzyloxyacetic acid 95%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: 2-(Benzyloxy)acetic acid

427080010 427080050
ACRO427080010EA 28.9 EUR
ACRO427080010 ACRO427080050
Benzyloxyacetic acid 95%
Benzyloxyacetic acid
Kaava: C₉H₁₀O₃
Kiehumispiste: 137…139 °C (,6 mmHg)
Tiheys: 1,162 g/cm³
Leimahduspiste: 113 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00274211
CAS-numero: 30379-55-6

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
HPLC ≥94 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR