2-Indancarboxylic acid 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

426360010 426360050
ACRO426360010EA 142 EUR
ACRO426360010 ACRO426360050
2-Indancarboxylic acid 98%
2-Indancarboxylic acid
Kaava: C₁₀H₁₀O₂
Sulamispiste: 128…132 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00085095
CAS-numero: 25177-85-9

Order Now

Testitulosten erittely

Appearance White to cream to brown Crystalline powder
Infrared spectrum Conforms
Melting point 128 to 132 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure
Sulfated ash ≤0.1 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤200 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR