1,4-Diiodoperfluorobutane 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

426320050
ACRO426320050EA 119 EUR
ACRO426320050
1,4-Diiodoperfluorobutane 98%
1,4-Diiodoperfluorobutane
Kaava: C₄F₈I₂
Kiehumispiste: 150 °C (1013 hPa)
Sulamispiste: –9 °C
Tiheys: 2,474 g/cm³
Storage Temperature: Jääkaappi
MDL numero: MFCD00042263
CAS-numero: 375-50-8

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow or pink Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4320 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR