Isoindoline 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

426280010 426280050
ACRO426280010EA 101 EUR
ACRO426280010 ACRO426280050
Isoindoline 97%
Isoindoline
Kaava: C₈H₉N
Kiehumispiste: 221 °C (760 mmHg)
Sulamispiste: 17 °C
Tiheys: 1.05 g/cm³
Leimahduspiste: 94 °C
Storage Temperature: Pakastin
MDL numero: MFCD00605324
CAS-numero: 496-12-8

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow to pink to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5670 to 1.5710 (20°C, 589 nm)
GC ≥96.0 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR