Metyylipunainen ACS

Toimittaja: Thermo Fisher Scientific

423970100 423970250 423971000
ACRO423970100EA 34.2 EUR
ACRO423970100 ACRO423970250 ACRO423971000
Metyylipunainen ACS
Metyylipunainen
Kaava: C₁₅H₁₅N₃O₂
Sulamispiste: 178…182 °C
Tiheys: 1,31 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002425
CAS-numero: 493-52-7
EINECS: 207-776-1
Merck Index: 12,06199

Order Now

Testitulosten erittely

Appearance Dark red to brown to purple Powder
Infrared spectrum Conforms
Melting point 179 to 182 °C
Visual transition interval From pH 4.2 (pink) to pH 6.2 (yellow)
Clarity of alcohol solution Passes test
Specific optical rotation +52.0° to +62.0° (20 °C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR