Hydroksi(nitroso)amino)bentsoliammoniumsuola

Toimittaja: Thermo Fisher Scientific
Vaara

423601000 423605000 423600250
ACRO423601000EA 74.1 EUR
ACRO423601000 ACRO423605000 ACRO423600250
Hydroksi(nitroso)amino)bentsoliammoniumsuola
Hydroksi(nitroso)amino)bentsoliammoniumsuola
Kaava: C₆H₉N₃O₂
Sulamispiste: 150…155 °C (haj. lämpötila)
Storage Temperature: Jääkaappi
MDL numero: MFCD00078422
CAS-numero: 135-20-6
EINECS: 205-183-2
UN: 2811
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance White to yellow-brown Crystalline powder or flakes
Infrared spectrum Conforms
Additional info package usually contains small pouches of ammonium carbonate as preservative
Solubility (5 % in water) Passes test
Residue after ignition ≤0.05 %
Suitability (for precip. of Fe, etc.) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR