Sinkkioksidi ACS

Toimittaja: Thermo Fisher Scientific
Varoitus

422740010 422740051 422741000 422745000
ACRO422740010EA 103 EUR
ACRO422740010 ACRO422740051 ACRO422741000 ACRO422745000
Sinkkioksidi ACS
Sinkkioksidi
Kaava: ZnO
Sulamispiste: 1975 °C
Tiheys: 5,6 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011300
CAS-numero: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Titration Complexometric ≥99.0 %
Alkalinity passes test
Chloride (Cl) ≤10 ppm
Nitrate (NO3) ≤30 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤50 ppm
Magnesium (Mg) ≤50 ppm
Manganese (Mn) ≤5 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤500 ppm
Sulfur compounds (as SO4) ≤0.01 %
Insoluble matter ≤0.01 % (in dilute H2SO4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR