Sodium polyanetholesulphonate

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: Polyanetholesulphonic acid sodium salt

419630250 419631000 419635000
ACRO419630250EA 179 EUR
ACRO419630250 ACRO419631000 ACRO419635000
Sodium polyanetholesulphonate
Sodium polyanetholesulphonate

CAS No.: 55963-78-5

Kaava: (C₁₀H₁₁O₄SNa)n
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00148427
CAS-numero: 55963-78-5

Order Now

Testitulosten erittely

Appearance White to yellow to beige to tan Powder or crystals
Infrared spectrum Conforms
pH 5.0 to 7.0 (1 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR