Hopeanitraatti 99+% ACS

Toimittaja: Thermo Fisher Scientific
Vaara

419350500 419351000 419352500 419355000 419350025
ACRO419350500EA 277 EUR
ACRO419350500 ACRO419351000 ACRO419352500 ACRO419355000 ACRO419350025
Hopeanitraatti 99+% ACS
Hopeanitraatti
Kaava: AgNO₃
Kiehumispiste: 444 °C (760 mmHg)
Sulamispiste: 212 °C
Tiheys: 4,35 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003414
CAS-numero: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White Crystalline powder or crystals
Free acid Passes test
Titration ≥99.0 %
Clarity of solution Passes test
Chloride (Cl) ≤5 ppm
Sulfate (SO4) ≤20 ppm
Copper (Cu) ≤2 ppm
Iron (Fe) ≤2 ppm
Lead (Pb) ≤10 ppm
Subst not pptd by HCl ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR