Fuksiini emäksinen 88%, pure

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline

419141000 419145000 419140025
ACRO419141000EA 88.3 EUR
ACRO419141000 ACRO419145000 ACRO419140025
Fuksiini emäksinen 88%, pure
Fuksiini emäksinen
Kaava: C₂₀H₂₀ClN₃
Sulamispiste: 250 °C (haj. lämpötila)
Tiheys: 1,22 g/cm³ (20 °C)
Leimahduspiste: 200 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00012569
CAS-numero: 632-99-5
EINECS: 211-189-6
Merck Index: 12,05686

Order Now

Testitulosten erittely

Appearance Green Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (105°C)
Dye content ≥83 % (on dry substance)
UV Lambda max 549 to 555 nm (in 50:50 IMS:water)
Absorption Ratio P-15/P+15 1.1 to 1.4
Specific absorbance A (1%/1cm) ≥1800 (at lambda max.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR