1-Pyrrolidinecarbonyl chloride 97%

Toimittaja: Thermo Fisher Scientific
Vaara

418650250 418650050
ACRO418650250EA 277 EUR
ACRO418650250 ACRO418650050
1-Pyrrolidinecarbonyl chloride 97%
1-Pyrrolidinecarbonyl chloride

CAS No.: 1192-63-8

Kaava: C₅H₈ClNO
Kiehumispiste: 75 °C (1 mmHg)
Tiheys: 1,209 g/cm³
Leimahduspiste: 98 °C
Storage Temperature: Pakastin
MDL numero: MFCD00051321
CAS-numero: 1192-63-8
UN: 3265
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4910 to 1.4970 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR