Kaliumjodaatti 99.4+% ACS

Toimittaja: Thermo Fisher Scientific
Vaara

418241000 418245000 418240025
ACRO418241000EA 61.7 EUR
ACRO418241000 ACRO418245000 ACRO418240025
Kaliumjodaatti 99.4+% ACS
Kaliumjodaatti
Kaava: KlO₃
Sulamispiste: 560 °C
Tiheys: 3,93 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011406
CAS-numero: 7758-05-6
EINECS: 231-831-9
UN: 1479
ADR: 5.1,II
Merck Index: 13,07726

Order Now

Testitulosten erittely

Appearance White Powder or crystalline powder
Titration Iodometric 99.4 to 100.4 %
pH 5.0 to 8.0 (5% soln. at 25°C)
Chloride and Bromide ≤0.01 % (as Cl)
Iodide (I) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Sodium (Na) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %
Nitrogen compounds (as N) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR