Sodium palmitate 98%

Toimittaja: Thermo Fisher Scientific

416700250 416700050 416701000
ACRO416700250EA 390 EUR
ACRO416700250 ACRO416700050 ACRO416701000
Sodium palmitate 98%
Sodium palmitate

CAS No.: 408-35-5

Kaava: C₁₆H₃₂O₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002749
CAS-numero: 408-35-5

Order Now

Testitulosten erittely

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (110°C, 4 hrs)
Titration with HClO4 97.5 to 102.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR