Naphthenic acid pract.

Toimittaja: Thermo Fisher Scientific
Varoitus

415281000 415285000 415280025
ACRO415281000EA 21.5 EUR
ACRO415281000 ACRO415285000 ACRO415280025
Naphthenic acid pract.
Naphthenic acid

CAS No.: 1338-24-5

Kiehumispiste: 140…200 °C (760 mmHg)
Tiheys: 0,91 g/cm³
Leimahduspiste: 149 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00147696
CAS-numero: 1338-24-5

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Density (20°C) 910 to 940 g/l
Viscosity ≤5 St (at 25°C)
Acid value 230 to 260 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR