Lanthanum(III) chloride hydrate ACS

Toimittaja: Thermo Fisher Scientific
Vaara

413010500
ACRO413010500EA 58.6 EUR
ACRO413010500
Lanthanum(III) chloride hydrate ACS
Lanthanum(III) chloride hydrate
Kaava: Cl₃La·H₂O
Kiehumispiste: ∼860 °C (1013 hPa)
Sulamispiste: 96 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149753
CAS-numero: 20211-76-1
EINECS: 233-237-5
UN: 1759

Order Now

Testitulosten erittely

Appearance White Adhering crystalline powder or crystals
Assay 64.5 to 70.0 % (LaCl3)
Calcium (Ca) ≤10 ppm
Magnesium (Mg) ≤10 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR