Lanoliini, vedetön USP

Toimittaja: Thermo Fisher Scientific

413005000 413000025
ACRO413005000EA 84.5 EUR
ACRO413005000 ACRO413000025
Lanoliini, vedetön USP
Lanoliini
Sulamispiste: 38…40 °C
Tiheys: 0,932 to 0.945 g/cm³ (15 °C)
Leimahduspiste: 209 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081740
CAS-numero: 8006-54-0
EINECS: 232-348-6
Merck Index: 12,05371

Order Now

Testitulosten erittely

Appearance Yellow Wax
Infrared spectrum Conforms
Melting point 38 to 44 °C
Water ≤0.25 %
Additional info Petrolatum: passes test
Antioxidant ≤0.02 % (BHT)
Residue after ignition ≤0.1 %
Acidity ≤2.0 ml (ml 0.1 N NaOH)
Alkalinity Passes test
Chloride (Cl) ≤0.035 %
Iodine value 18 to 36
Ammonia (NH3) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR