2-Jodifenoli 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

412480250 412480050 412481000
ACRO412480250EA 91.6 EUR
ACRO412480250 ACRO412480050 ACRO412481000
2-Jodifenoli 98%
2-Jodifenoli
Kaava: C₆H₅IO
MW: 220.01 g/mol
Kiehumispiste: 186…187 °C (160 mmHg)
Sulamispiste: 37…43 °C
Leimahduspiste: >110 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013963
CAS-numero: 533-58-4
EINECS: 208-569-9
Merck Index: 13,05057

Order Now

Testitulosten erittely

Appearance White to brown-red to brown Crystalline powder or low melting mass
Infrared spectrum Conforms
Melting point 37 to 43 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR