4-Jodibentsyylikloridi 98%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: p-Iodobenzoyl chloride

412390250 412390050
ACRO412390250EA 175 EUR
ACRO412390250 ACRO412390050
4-Jodibentsyylikloridi 98%
4-Jodibentsyylikloridi
Kaava: C₇H₄ClIO
MW: 266.47 g/mol
Kiehumispiste: 100 °C (0.7 mmHg)
Sulamispiste: 62…66 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001058
CAS-numero: 1711-02-0
EINECS: 216-974-7
UN: 3261
ADR: 8 III

Order Now

Testitulosten erittely

Appearance Light yellow to beige to light brown Crystalline powder and/or chunks or solid
Infrared spectrum Conforms
Melting point 62 to 66 °C
GC ≥97.5 %
Acidity ≤0.0002 meq/g
Peroxides ≤0.002 % (as H2O2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR