Acetoin dimer 96%

Toimittaja: Thermo Fisher Scientific
Varoitus

411951000 411955000
ACRO411951000EA 94.3 EUR
ACRO411951000 ACRO411955000
Acetoin dimer 96%
Acetoin dimer
Kaava: C₈H₁₆O₄
Kiehumispiste: 148 °C (1013 hPa)
Sulamispiste: 90…91 °C
Tiheys: 0,998 g/cm³ (20 °C)
Storage Temperature: Jääkaappi
MDL numero: MFCD00004521
CAS-numero: 23147-57-1
EINECS: 208-174-1
UN: 1325
ADR: 4.1,III
Merck Index: 13,00065

Order Now

Testitulosten erittely

Appearance White to yellow Powder or crystals
Infrared spectrum Conforms
GC ≥95 %
Additional info Polymerization can be reversed by heating at 120°C
Specific gravity (25°C) 0.995 to 1.019


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR