EGTA (Etyleeniglykolibis(2-aminoetyylieetteri)-N,N,N',N'-tetraetikkahappo) 98%

Toimittaja: Thermo Fisher Scientific

409910250 409911000 409915000
ACRO409910250EA 124 EUR
ACRO409910250 ACRO409911000 ACRO409915000
EGTA (Etyleeniglykolibis(2-aminoetyylieetteri)-N,N,N',N'-tetraetikkahappo) 98%
EGTA (Etyleeniglykolibis(2-aminoetyylieetteri)-N,N,N',N'-tetraetikkahappo)
Kaava: C₁₄H₂₄N₂O₁₀
Leimahduspiste: >200 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004291
CAS-numero: 67-42-5
EINECS: 200-651-2
Merck Index: 13,03560

Order Now

Testitulosten erittely

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 240 to 244 °C
Titration excess NaOH ≥97.5 %
HPLC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR