2-Ethylbutyryl chloride 99%

Toimittaja: Thermo Fisher Scientific
Vaara

409821000 409820050 409820250
ACRO409821000EA 240 EUR
ACRO409821000 ACRO409820050 ACRO409820250
2-Ethylbutyryl chloride 99%
2-Ethylbutyryl chloride
Kaava: C₆H₁₁ClO
Leimahduspiste: 33 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00018812
CAS-numero: 2736-40-5
EINECS: 220-359-9
UN: 2920
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4250 (20°C, 589 nm)
Titration RCOCl ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR