Diphenylphosphoryl azide 98%

Toimittaja: Thermo Fisher Scientific
Vaara

409040250 409041000 409045000
ACRO409040250EA 103 EUR
ACRO409040250 ACRO409041000 ACRO409045000
Diphenylphosphoryl azide 98%
Diphenylphosphoryl azide
Kaava: C₁₂H₁₀N₃O₃P
Kiehumispiste: 157 °C (,2 mmHg)
Tiheys: 1,273 g/cm³
Leimahduspiste: 112 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00001987
CAS-numero: 26386-88-9
EINECS: 247-644-0
UN: 3278
ADR: 6.1,II

Order Now

Testitulosten erittely

Appearance Clear yellow to amber Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5520 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR