Dimethylthiocarbamoyl chloride 99%

Toimittaja: Thermo Fisher Scientific
Vaara

408610250 408611000
ACRO408610250EA 85.4 EUR
ACRO408610250 ACRO408611000
Dimethylthiocarbamoyl chloride 99%
Dimethylthiocarbamoyl chloride

Appearance: Yellow Crystals or low melting crystalline solid

Kaava: C₃H₆ClNS
Kiehumispiste: 63…65 °C (,2mbar)
Sulamispiste: 42…43 °C
Leimahduspiste: 98 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004919
CAS-numero: 16420-13-6
UN: 3261
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Yellow Crystals or low melting crystalline solid
Infrared spectrum Conforms
GC ≥98.5 %
Appearance of solution (10% in CHCl3) clear faint yellow to yellow


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR