Diethyl iminodiacetate 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

407320250 407321000 407320050
ACRO407320250EA 151 EUR
ACRO407320250 ACRO407321000 ACRO407320050
Diethyl iminodiacetate 98%
Diethyl iminodiacetate

Appearance: Clear colorless to yellow Liquid

Kaava: C₈H₁₅NO₄
Kiehumispiste: 248 °C (760 mmHg)
Tiheys: 1,08 g/cm³
Leimahduspiste: 104 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00041925
CAS-numero: 6290-05-7

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4350 to 1.4380 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR