Di-tert-butyl malonate 98%

Toimittaja: Thermo Fisher Scientific

406620100 406620500
ACRO406620100EA 59.7 EUR
ACRO406620100 ACRO406620500
Di-tert-butyl malonate 98%
Di-tert-butyl malonate
Kaava: C₁₁H₂₀O₄
Kiehumispiste: 110…111 °C (22 mmHg)
Sulamispiste: –7 °C
Tiheys: 0,966 g/cm³
Leimahduspiste: 88 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00008810
CAS-numero: 541-16-2
EINECS: 208-769-6

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4175 to 1.4195 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR