1,3-Dibromiasetoni 70% tech.

Toimittaja: Thermo Fisher Scientific
Varoitus

406550250
ACRO406550250EA 377 EUR
ACRO406550250
1,3-Dibromiasetoni 70% tech.
1,3-Dibromiasetoni
Kaava: BrH₂CC(O)CH₂Br
Kiehumispiste: 95 °C (20 mmHg)
Sulamispiste: 28 °C
Tiheys: 2,167 g/cm³ (18 °C)
MDL numero: MFCD00013540
CAS-numero: 816-39-7
EINECS: 212-430-8
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Yellow to dark brown Liquid
Infrared spectrum Conforms
GC ≥99.9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR