Carbohydrazide 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

403970250 403971000 403975000
ACRO403970250EA 50.9 EUR
ACRO403970250 ACRO403971000 ACRO403975000
Carbohydrazide 97%
Carbohydrazide
Kaava: CH₆N₄O
MW: 90.09 g/mol
Sulamispiste: >153 °C
Tiheys: 1.02 g/cm³
Storage Temperature: Jääkaappi
MDL numero: MFCD00007591
CAS-numero: 497-18-7
EINECS: 207-837-2
UN: 3077
ADR: 9 III

Order Now

Testitulosten erittely

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 153 to 158 °C
Titration with KIO3 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR