Kalsiumoksalaatti-H2O 98%, extra pure

Toimittaja: Thermo Fisher Scientific
Varoitus

403880050 403885000 403880025
ACRO403880050EA 35.1 EUR
ACRO403880050 ACRO403885000 ACRO403880025
Kalsiumoksalaatti-H2O 98%, extra pure
Kalsiumoksalaatti-H2O
Kaava: C₂CaO₄·1H₂O
Sulamispiste: 200 °C
Tiheys: 2,2 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00150039
CAS-numero: 5794-28-5
EINECS: 209-260-1
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤13 % (1 g, 200°C, 3 h)
Particle size ≥95 % (Passing through 45 µm)
Apparent density 0.3 to 0.5 g/cm3
Titration ≥97.5 % w/w (as Oxalate) On dry substance
Iron (Fe) ≤200 ppm
Insoluble matter ≤0.3 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR