Kadmiumsulfaatti-8/3 H2O, kiteinen ACS

Toimittaja: Thermo Fisher Scientific
Vaara

403765000 403760025 403761000
ACRO403765000EA 192 EUR
ACRO403765000 ACRO403760025 ACRO403761000
Kadmiumsulfaatti-8/3 H2O, kiteinen ACS
Kadmiumsulfaatti-8/3 H2O
Kaava: 3CdSO₄·8H₂O
Sulamispiste: 41 °C
Tiheys: 3,79 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02253000
CAS-numero: 7790-84-3
EINECS: 233-331-6
UN: 2570
ADR: 6.1,III
Merck Index: 13,01627

Order Now

Testitulosten erittely

Appearance White Crystals
Titration Complexometric 98.0 to 102.0 %
Chloride (Cl) ≤0.001 %
Nitrate and Nitrite (as NO3) ≤0.003 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.002 %
Iron (Fe) ≤0.001 %
Lead (Pb) ≤0.003 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.02 %
Zinc (Zn) ≤0.05 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR