Bis(2-ethylhexyl)phosphate 95%

Toimittaja: Thermo Fisher Scientific
Vaara

402470010 402470050
ACRO402470010EA 266 EUR
ACRO402470010 ACRO402470050
Bis(2-ethylhexyl)phosphate 95%
Bis(2-ethylhexyl)phosphate
Kaava: C₁₆H₃₅O₄P
Kiehumispiste: 48 °C (12 mmHg)
Sulamispiste: –60 °C
Tiheys: 0,97 g/cm³
Leimahduspiste: 137 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009492
CAS-numero: 298-07-7
EINECS: 206-056-4
UN: 1902
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4440 (20°C, 589 nm)
Titration with NaOH ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR