Ethyl hydrogen malonate 95%

Toimittaja: Thermo Fisher Scientific
Varoitus

399900050 399900250 399902500
ACRO399900050EA 48.9 EUR
ACRO399900050 ACRO399900250 ACRO399902500
Ethyl hydrogen malonate 95%
Ethyl hydrogen malonate

Appearance: Clear colorless to light yellow Liquid

Kaava: C₅H₈O₄
Kiehumispiste: 106,5 °C (3 mmHg)
Tiheys: 1,119 g/cm³
Storage Temperature: Jääkaappi
MDL numero: MFCD00020490
CAS-numero: 1071-46-1

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Titration with NaOH ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR