1-Boc-4-(hydroxymethyl)piperidine 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

397720010 397720050
ACRO397720010EA 19.1 EUR
ACRO397720010 ACRO397720050
1-Boc-4-(hydroxymethyl)piperidine 97%
1-Boc-4-(hydroxymethyl)piperidine
Kaava: C₁₁H₂₁NO₃
Sulamispiste: 78…82 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02094488
CAS-numero: 123855-51-6

Order Now

Testitulosten erittely

Appearance White to brown Powder
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR