Magnesium ethanolate 98%

Toimittaja: Thermo Fisher Scientific
Vaara

Synonyms: Magnesium ethoxide, Magnesium ethylate

397470050 397471000 397475000
ACRO397470050EA 44.4 EUR
ACRO397470050 ACRO397471000 ACRO397475000
Magnesium ethanolate 98%
Magnesium ethanolate
Kaava: C₄H₁₀MgO₂
Sulamispiste: 270 °C (haj. lämpötila)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009060
CAS-numero: 2414-98-4
UN: 3205
ADR: 4.2,II

Order Now

Testitulosten erittely

Appearance White to light grey Powder
Infrared spectrum Conforms
Additional info May contain up to 2.5% Mg(OH)2 and/or MgCO3
Titration with HCl 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR