1-Boc-3-piperidone 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

397000050 397000250 397001000
ACRO397000050EA 99.1 EUR
ACRO397000050 ACRO397000250 ACRO397001000
1-Boc-3-piperidone 97%
1-Boc-3-piperidone
Kaava: C₁₀H₁₇NO₃
Sulamispiste: 37…39 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01631193
CAS-numero: 98977-36-7

Order Now

Testitulosten erittely

Appearance White to yellow to orange Solid or liquid
Infrared spectrum Conforms
GC ≥96.0 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR