(±)-1-Phenyl-2-propyn-1-ol 98+%

Toimittaja: Thermo Fisher Scientific
Varoitus

392760010 392760050 392760250
ACRO392760010EA 46.6 EUR
ACRO392760010 ACRO392760050 ACRO392760250
(±)-1-Phenyl-2-propyn-1-ol 98+%
(±)-1-Phenyl-2-propyn-1-ol
Kaava: C₉H₈O
Sulamispiste: 22 °C
Leimahduspiste: 99 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00021860
CAS-numero: 4187-87-5

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5530 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR