Indole-5-carbaldehyd 98+%

Toimittaja: Thermo Fisher Scientific
Varoitus

391630010 391630050 391630250
ACRO391630010EA 44.4 EUR
ACRO391630010 ACRO391630050 ACRO391630250
Indole-5-carbaldehyd 98+%
Indole-5-carbaldehyd
Kaava: C₉H₇NO
Sulamispiste: 100…103 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02093664
CAS-numero: 1196-69-6

Order Now

Testitulosten erittely

Appearance Orange to tan Powder
Infrared spectrum Conforms
Melting point 98 to 103 °C
HPLC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR