trans-Bis(tributylstannyl)ethylene 95%

Toimittaja: Thermo Fisher Scientific
Vaara

388670010 388670050
ACRO388670010EA 87.1 EUR
ACRO388670010 ACRO388670050
trans-Bis(tributylstannyl)ethylene 95%
trans-Bis(tributylstannyl)ethylene
Kaava: C₂₆H₅₆Sn₂
Storage Temperature: Jääkaappi
MDL numero: MFCD01631299
CAS-numero: 14275-61-7
UN: 2788
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5000 to 1.5030 (20°C, 589 nm)
NMR ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR