Natriumkloridi ∼26% in water, pure

Toimittaja: Thermo Fisher Scientific

387640010 387640025
ACRO387640010EA 29.6 EUR
ACRO387640010 ACRO387640025
Natriumkloridi ∼26% in water, pure
Natriumkloridi
Kaava: NaCl
Tiheys: 1,202 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003477
CAS-numero: 7647-14-5

Order Now

Testitulosten erittely

Appearance Clear colorless Solution
Titration Argentometric 25 to 28 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR