Titaani(IV)oksidi, Aeroxide® P25

Toimittaja: Thermo Fisher Scientific

384290500 384292500 384290010
ACRO384290500EA 82.2 EUR
ACRO384290500 ACRO384292500 ACRO384290010
Titaani(IV)oksidi, Aeroxide® P25
Titaani(IV)oksidi
Kaava: TiO₂
Kiehumispiste: 2900 °C (760 mmHg)
Sulamispiste: 1855 °C
Tiheys: 4,23 g/cm³
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011269
CAS-numero: 13463-67-7
EINECS: 236-675-5
Merck Index: 13,09549

Order Now

Testitulosten erittely

Appearance White Powder
Assay ≥99.5 % (on ignited substance) Typical
Specific surface area 35 to 65 m²/g
pH 3.5 to 4.5 (4 % dispersion)
Acidity (HCl) ≤0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR