(R)-(+)-3-Aminoquinuclidine dihydrochloride 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

382890010
ACRO382890010EA 78.5 EUR
ACRO382890010
(R)-(+)-3-Aminoquinuclidine dihydrochloride 98%
(R)-(+)-3-Aminoquinuclidine dihydrochloride
Kaava: C₇H₁₆Cl₂N₂
Sulamispiste: >300 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00191752
CAS-numero: 123536-14-1

Order Now

Testitulosten erittely

Appearance White to beige Powder
Infrared spectrum Conforms
Specific optical rotation +22° to +26° (20°C, 589 nm) (C=1, Water)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR