N-(tert-Butoxycarbonyl)-4-aminobutyric acid 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

376570010 376570050 376570250
ACRO376570010EA 45.4 EUR
ACRO376570010 ACRO376570050 ACRO376570250
N-(tert-Butoxycarbonyl)-4-aminobutyric acid 98%
N-(tert-Butoxycarbonyl)-4-aminobutyric acid

CAS No.: 57294-38-9

Kaava: C₉H₁₇NO₄
Kiehumispiste: 78…82 °C (,2 mmHg)
Sulamispiste: 55 °C
Leimahduspiste: >110 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00037313
CAS-numero: 57294-38-9

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 54 to 60 °C
TLC ≥97.5 %
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR