Ethyl isodehydroacetate 95%

Toimittaja: Thermo Fisher Scientific

373900250
ACRO373900250EA 101 EUR
ACRO373900250
Ethyl isodehydroacetate 95%
Ethyl isodehydroacetate
Kaava: C₁₀H₁₂O₄
Kiehumispiste: 290…295 °C (760 mmHg)
Sulamispiste: 17…18 °C
Tiheys: 1,167 g/cm³
Leimahduspiste: >110 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006643
CAS-numero: 3385-34-0
EINECS: 222-195-3

Order Now

Testitulosten erittely

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5135 to 1.5165 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR