2-Phenoxyaniline 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

373160250
ACRO373160250EA 62.6 EUR
ACRO373160250
2-Phenoxyaniline 98%
2-Phenoxyaniline
Kaava: C₁₂H₁₁NO
Kiehumispiste: 170 °C (18 mmHg)
Sulamispiste: 46…50 °C
Leimahduspiste: 113 °C
MDL numero: MFCD00035765
CAS-numero: 2688-84-8
EINECS: 220-254-8

Order Now

Testitulosten erittely

Appearance Brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 45 to 49 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR