2-Kinoliinikarbaldehydi 97%

Toimittaja: Thermo Fisher Scientific
Varoitus

370170050 370170250
ACRO370170050EA 86.2 EUR
ACRO370170050 ACRO370170250
2-Kinoliinikarbaldehydi 97%
2-Kinoliinikarbaldehydi
Kaava: C₁₀H₇NO
Sulamispiste: 66…71 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00075032
CAS-numero: 5470-96-2
EINECS: 226-804-3

Order Now

Testitulosten erittely

Appearance Yellow to orange to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 66 to 71 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR