Aminoguanidine hydrochloride 98%

Toimittaja: Thermo Fisher Scientific
Varoitus

368910250 368911000
ACRO368910250EA 68 EUR
ACRO368910250 ACRO368911000
Aminoguanidine hydrochloride 98%
Aminoguanidine hydrochloride
Kaava: CH₇ClN₄
Sulamispiste: >160 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00039074
CAS-numero: 1937-19-5
EINECS: 217-707-7

Order Now

Testitulosten erittely

Appearance White to off-white Crystals
Infrared spectrum Conforms
Melting point ≥160 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR