5-Bromo-2-chloropyrimidine 96%

Toimittaja: Thermo Fisher Scientific
Varoitus

368482500 368480050
ACRO368482500EA 31.2 EUR
ACRO368482500 ACRO368480050
5-Bromo-2-chloropyrimidine 96%
5-Bromo-2-chloropyrimidine
Kaava: C₄H₂BrClN₂
Sulamispiste: 78…80 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00483232
CAS-numero: 32779-36-5

Order Now

Testitulosten erittely

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥95 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR