Ethylene diacrylate 95% stabiloitu

Toimittaja: Thermo Fisher Scientific
Vaara

366970050 366970250
ACRO366970050EA 171 EUR
ACRO366970050 ACRO366970250
Ethylene diacrylate 95% stabiloitu
Ethylene diacrylate

Appearance: Clear colorless to light yellow Liquid

Kaava: C₈H₁₀O₄
Kiehumispiste: 62…65 °C (2 mmHg)
Tiheys: 1,04 g/cm³
Leimahduspiste: 101 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00008629
CAS-numero: 2274-11-5
UN: 2810
ADR: 6.1,III

Order Now

Testitulosten erittely

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4560 (20°C, 589 nm)
GC ≥94 %
Stabilizer 100 to 400 ppm MEHQ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR