Coconut oil, pure, refined

Toimittaja: Thermo Fisher Scientific

365471000 365475000
ACRO365471000EA 32.2 EUR
ACRO365471000 ACRO365475000
Coconut oil, pure, refined
Coconut oil
Sulamispiste: 23…26 °C
Leimahduspiste: >200 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00130817
CAS-numero: 8001-31-8
EINECS: 232-282-8

Order Now

Testitulosten erittely

Appearance Colorless to white to light yellow Oily liquid or solid
Acid value ≤0.5 mg KOH/g
Iodine value 7 to 12 g I2/100 g
Peroxide value ≤5 meq O2/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR