Benzyltrimethylammonium bromide 98+%

Toimittaja: Thermo Fisher Scientific
Varoitus

Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium bromide

364860250 364861000
ACRO364860250EA 26 EUR
ACRO364860250 ACRO364861000
Benzyltrimethylammonium bromide 98+%
Benzyltrimethylammonium bromide
Kaava: C₁₀H₁₆BrN
Sulamispiste: 228…234 °C
MDL numero: MFCD00011780
CAS-numero: 5350-41-4
EINECS: 226-320-2
UN: 2811

Order Now

Testitulosten erittely

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Melting point 224 to 234 °C
Titration Argentometric ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR