Silikageeli C8-RP 12%C, ∼ 1.2 mmol/g 40-63 µm

Toimittaja: Thermo Fisher Scientific

360140500 360142500
ACRO360140500EA 226 EUR
ACRO360140500 ACRO360142500
Silikageeli C8-RP 12%C, ∼ 1.2 mmol/g 40-63 µm
Silikageeli
Kaava: SiO₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00148339
CAS-numero: 7631-86-9

Order Now

Testitulosten erittely

Appearance White to off-white Powder
Particle size 40 to 63 µm
Elemental Analysis ≥11.6 % (C)
Loading ≥1.21 mmol/g
Volatile matter ≤4 % (110°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR